Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCH4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191639
Description
44259 Polyclonal specifically detects 44259 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MARCH4 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q9P2E8 | |
43163 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:WKVLNYDKTKDLEDQKAGGRTNPRTSSSTQANIPSSEEETAGTPAPEQGPAQAAGHPSGPLSHHHCAYTILHILSHLRPHEQRSP | |
0.1 mL | |
Zinc Finger | |
57574 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 6.3.2.-, KIAA1399E3 ubiquitin-protein ligase MARCH4, MARCH-IVMGC104908, membrane-associated ring finger (C3HC4) 4, Membrane-associated RING finger protein 4, Membrane-associated RING-CH protein IV, RNF174RING finger protein 174 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction