Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCKS Mouse anti-Bovine, Human, Clone: 2F12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05337
Description
Anti-MARCKS Antibody, clone 2F12 detects level of MARCKS and has been published and validated for use in immunoprecipitation and western blotting.Specifications
MARCKS | |
Monoclonal | |
Unconjugated | |
NM_002356 | |
Mouse | |
Ascites | |
Primary | |
−20°C for up to 6 months. | |
IgG1 |
Immunoprecipitation, Western Blot | |
2F12 | |
mouse ascites containing 0.05% sodium azide | |
MARCKS, 80K-L, MACS, PRKCSL, FLJ14368, phosphomyristin, MRACKS, PKCSL, FLJ90045 | |
Purified recombinant human MARCKS; Epitope mapped to the carboxyl-terminal. | |
200 μL | |
Bovine, Human | |
Ascites |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only