Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methionyl-tRNA synthetase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MARS protein.

Manufacturer:  Abnova Corporation H00004141B01

Catalog No. 89-005-733


Add to cart

Description

Description

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene belongs to the class I family of tRNA synthetases. [provided by RefSeq

Sequence: MRLFVSDGVPGCLPVLAAAGRARGRAEVLISTVGPEDCVVPFLTRPKVPVLQLDSGNYLFSTSAICRYFFLLSGWEQDDLTNQWLEWEATELQPALSAALYYLVVQGKKGEDVLGSVRRALTHIDHSLSRQNCPFLAGETESLADIVLWGALYPLLQDPAYLPEELSALHSWFQTLSTQEPCQRAAETVLKQQGVLALRPYLQKQPQPSPAEGRAVTNEPEEEELATLSEEEIAMAVTAWEKGLESLPPLRPQQNPVLPVAGERNVLITSALPYVNNVPHLGNIIGCVLSADVFARYSRLRQWNTLYLCGTDEYGTATETKALEEGLTPQEICDKYHIIHADIYRWFNISFDIFGRTTTPQQTKITQDIFQQLLKRGFVLQDTVEQLRCEHCARFLADRFVEGVCPFCGYEEARGDQCDKCGKLINAVELKKPQCKVCRSCPVVQSSQHLFLDLPKLEKRLEEWLGRTLPGSDWTPNAQFITRSWLRDGLKPRCITRDLKWGTPVPLEGFEDKVFYVWFDATIGYLSITANYTDQWERWWKNPEQVDLYQFMAKDNVPFHSLVFPCSALGAEDNYTLVSHLIATEYLNYEDGKFSKSRGVGVFGDMAQDTGIPADIWRFYLLYIRPEGQDSAFSWTDLLLKNNSELLNNLGNFINRAGMFVSKFFGGYVPEMVLTPDDQRLLAHVTLELQHYHQLLEKVRIRDALRSILTISRHGNQYIQVNEPWKRIKGSEADRQRAGTVTGLAVNIAALLSVMLQPYMPTVSATIQAQLQLPPPACSILLTNFLCTLPAGHQIGTVSPLFQKLENDQIESLRQRFGGGQAKTSPKPAVVETVTTAKPQQIQALMDEVTKQGNIVRELKAQKADKNEVAAEVAKLLDLKKQLAVAEGKPPEAPKGKKKK
Specifications

Specifications

methionyl-tRNA synthetase
Unconjugated
No additive
NM_004990.2
FLJ35667|METRS|MTRNS
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4141
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human MARS protein.
MARS
Serum
MARS
MARS (NP_004981.2, 1 a.a. ~ 900 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit