Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mas, DyLight 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178444V

Catalog No. NBP178444V


Add to cart

Description

Description

Mas Polyclonal antibody specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mas
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
MAS1
A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, monkey, rat, and mouse. Rabbit reactivity reported in the scientific literature (PMID: 23701246).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
P04201
MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas
Rabbit
IgG
0.1 ml
Cancer, GPCR, Neuroscience
Primary
4142
Human, Mouse, Primate, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only