Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mas, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP179143

Catalog No. NBP179143

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Mas
1 mg/mL
Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100
P04201
MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas
Rabbit
IgG
25ug
GPCR, Neuroscience
Primary
4142
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.7) with 0.01% Sodium Azide
Affinity Purified
MAS1
Synthetic peptide (aa sequence is considered to be commercially sensitive)
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
It detects MAS1 in Human samples.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only