Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mas, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$180.00 - $261.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen KLH conjugated synthetic peptide made to the C-terminus of human MAS1. [UniProt# P04201]
Antigen Mas
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NLS1531SS
View Documents
Novus Biologicals
NLS1531SS
0.025 ml
Each for $180.00
Add to cart
 
NLS1531
View Documents
Novus Biologicals
NLS1531
0.05 ml
Each for $261.50
Add to cart
 
Description

Description

Mas Polyclonal antibody specifically detects Mas in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Rabbit
KLH conjugated synthetic peptide made to the C-terminus of human MAS1. [UniProt# P04201]
Polyclonal
Primary
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
P04201
PBS (pH 7.7) with 0.01% Sodium Azide
IgG
Mas
Affinity Purified
RUO
Human and mouse. Immunogen sequence has 82% identity with rat.
4142
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit