Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Maspin, Clone: 3B8.2,
Supplier: MilliporeSigma™ MAB4035
Description

Specifications
Maspin | |
Monoclonal | |
Unconjugated | |
NM_002639.3 | |
Mouse | |
100 μg | |
Human | |
Purified |
Western Blot | |
3B8.2 | |
Protein A purified immunoglobulin. Liquid in 0.02MPhosphate with 0.15MNaCl, pH 7.6 and 0.1% sodium azide as a preservative. | |
SERPINB5, PI5, Maspin, maspin | |
Synthetic peptide corresponding to amino acids 112-130 of human Maspin. The immunogen sequence is 100% identical in mouse. | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction