Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCP11 Rat anti-Mouse, Clone: 451421, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524171

Catalog No. PIMA524171


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse (rm) MCP-1, rmMCP-6, or rmMCP-7 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

MCP11
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
Prss34
Mouse myeloma cell line NS0-derived recombit mouse Mast Cell Protease-11/Prss34 Met20-Ser318
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Immunoprecipitation, Western Blot
451421
Unconjugated
Prss34
MCP-11, MCPT11, Prss34
Rat
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
328780
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit