Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASTL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525934

Catalog No. PIPA525934

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Serine/threonine kinase that plays a key role in M phase by acting as a regulator of mitosis entry and maintece. Acts by promoting the inactivation of protein phosphatase 2A (PP2A) during M phase: does not directly inhibit PP2A but acts by mediating phosphorylation and subsequent activation of ARPP19 and ENSA at  ft.Ser-62 ft. and  ft.Ser-67 ft., respectively. ARPP19 and ENSA are phosphatase inhibitors that specifically inhibit the PPP2R2D (PR55-delta) subunit of PP2A. Inactivation of PP2A during M phase is essential to keep cyclin-B1-CDK1 activity high. Following DNA damage, it is also involved in checkpoint recovery by being inhibited. Phosphorylates histone protein in vitro; however such activity is unsure in vivo. May be involved in megakaryocyte differentiation.
Specifications

Specifications

MASTL
Polyclonal
PBS with 0.09% sodium azide
Q96GX5
GREATWALL, GW, GWL, MAST-L, RP11-85G18.2, THC2, hGWL, greatwall protein kinase, microtubule-associated serine/threonine-protein kinase-like, serine/threonine-protein kinase greatwall
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84930
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MASTL
Liquid
MASTL
human partial MASTL recombit protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit