Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAT2A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232283

Catalog No. NBP232283


Add to cart

Description

Description

MAT2A Polyclonal antibody specifically detects MAT2A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MAT2A
1.0 mg/mL
Western Blot 1:2000 - 1:10000
NP_005902.1
AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2
Rabbit
IgG
100 ul
Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction
Primary
4144
Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
MAT2A
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Methionine Adenosyltransferase II, Alpha using the numbering given in entry NP_005902.1 (GeneID 4144).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only