Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MATE1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136989

Catalog No. NBP136989


Add to cart

Description

Description

MATE1 Polyclonal antibody specifically detects MATE1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

MATE1
0.5 mg/mL
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:64000
NP_060712.2
hMATE-1, MATE-1, MATE1FLJ10847multidrug and toxin extrusion 1, MGC64822, multidrug and toxin extrusion protein 1, Solute carrier family 47 member 1, solute carrier family 47, member 1
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region (near C-Terminus).
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
SLC47A1
Peptide with sequence C-PEHPQDGAKLSRKQ corresponding to internal region (near C-Terminus) according to NP_060712.2.
Immunogen affinity purified
RUO
Primary
55244
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only