Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATH1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512259

Catalog No. PIPA512259

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ATOH1 belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47.
Specifications

Specifications

MATH1
Polyclonal
PBS with 0.09% sodium azide
Q92858
Protein atonal homolog 1; Helix-loop-helix protein hATH-1; ATOH1; ATH1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
474
Western Blot
Unconjugated
ATOH1
Liquid
ATOH1
KLH conjugated synthetic peptide between 78-109 amino acids from the N-terminal region of human MATH1/HATH1/ATOH1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit