Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATH1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131318

Catalog No. NBP131318


Add to cart

Description

Description

MATH1 Polyclonal antibody specifically detects MATH1 in Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

MATH1
0.75mg/mL
Immunocytochemistry/Immunofluorescence 1:100-1:1000
Q92858
ATH1protein atonal homolog 1, atonal homolog 1 (Drosophila), BHLHA14, bHLHa14Math1, Class A basic helix-loop-helix protein 14, hATH1, Helix-loop-helix protein hATH-1, MATH-1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Affinity Purified
ATOH1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MATH1. The exact sequence is proprietary.
38 kDa
0.1 ml
Cancer
Primary
474
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only