Learn More
MAVS/Cardif Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061043
Catalog No. 06-104-3MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-MAVS/Cardif Antibody, N-terminus is an antibody against MAVS/Cardif for use in western blotting and immunoprecipitation.
Specifications
Specifications
MAVS/Cardif
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
MAVS; VISA; IPS-1; IPS1
GST-tagged recombinant protein corresponding to the N-terminus of Mitochondrial antiviral-signaling protein.
100μL
Primary
NP_065797