Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAX, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA132194

Catalog No. PIPA132194

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown.

PA1-32194 detects MAX from human samples.PA1-32194 has been successfully used in Western blot applications.The PA1-32194 immunogen is: Peptide with sequence EEPQSRKKLRMEAS, from C-Terminus of the protein sequence according to NP_002373
Specifications

Specifications

MAX
Polyclonal
Unconjugated
MAX
Purified
MAX
Peptide with sequence EEPQSRKKLRMEAS, from C-Terminus of the protein sequence according to NP_002373
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P61244, P28574
MGC10775, MGC11225, MGC18164, MGC34679, MGC36767, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8, orf1
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
4149
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit