Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Maxi Potassium channel alpha, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136966

Catalog No. NBP136966


Add to cart

Description

Description

Maxi Potassium channel alpha Polyclonal antibody specifically detects Maxi Potassium channel alpha in Human, Mouse, Rat, Bovine samples. It is validated for Peptide ELISA.
Specifications

Specifications

Maxi Potassium channel alpha
0.5 mg/mL
Peptide ELISA Detection limit 1:2000
NP_001014797.1
bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1
Goat
IgG
0.1 mg
Potassium Channels
Primary
3778
Bovine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
KCNMA1
Peptide with sequence C-RESRDKQNRKEMVYR corresponding to C-Terminus according to NP_001014797.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize isoform a (NP_001014797.1) only.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only