Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MBD-R2, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 49940002
Description
MBD-R2 Polyclonal antibody specifically detects MBD-R2 in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
MBD-R2 | |
1 mg/mL | |
Western Blot 1:100-1:2000, Chromatin Immunoprecipitation 1:10-1:500 | |
Affinity Purified | |
Mbd-R2 | |
In vivo generated recombinant protein fragment. | |
Immunogen affinity purified | |
RUO | |
Primary | |
41498 | |
Drosophila |
ChIP assay, Western Blot | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
CG10042, CG17393, Dm Tam3, dMBD-R2, Dmel\CG10042, MBD, MBDR2, TAM3 | |
Rabbit | |
IgG | |
0.05 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Drosophila . |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only