Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MBD4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191189SS

 View more versions of this product

Catalog No. NBP191189SS


Add to cart

Description

Description

MBD4 Polyclonal antibody specifically detects MBD4 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

MBD4
1.11 mg/mL
Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative)
O95243
EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein
Rabbit
IgG
0.025 ml
Chromatin Research, DNA Repair, Epigenetics
Primary
8930
Human, Mouse (Negative)
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
Affinity Purified
MBD4
A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human. Does not react with mouse.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only