Learn More
MBIP Mouse anti-Human, Clone: 2E2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521956
Catalog No. PIMA521956
Add to cart
Description
Description
Peptide Sequence: MAAATEHNRP SSGDRNLERR CSPNLSREVL YEIFRSLHTL VGQLDLRDDV VKITIDWNKL QSLSAFQPAL LFSALEQHIL YLQPFLAKLQ SPIKEENTTA VEETGRTEMG NKNEVNDKFS IGDLQEEEKH KESDLRDVKK TQIHFDPEVV QIKAGKAEID RRISAFIERK QAEINENNVR EFCNVIDCNQ ENSCARTDAI FTPYPGFKSH VKVSRVVYTY GPQTRPEGIP GSGHKPNSML RDCGNQAVEE RLQNIEAHLR LQTGGPVPRD IYQRIKKLED KILELEGISP EYFQSVSFSG KRRKVQPPQQ NYSLAELDEK ISALKQALLR KSREAESMAT HHLP
Inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway.
Specifications
Specifications
MBIP
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q9NS73
MAP3K12-binding inhibitory protein 1, MAPK upstream kinase-binding inhibitory protein, MUK-binding inhibitory protein
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51562
ELISA, Immunocytochemistry, Immunofluorescence
2E2
Unconjugated
MBIP
Liquid
MBIP
MBIP (AAH05197, 1 a.a. ∽ 344 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human