Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBL2 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA518325

Catalog No. PIPA518325


Add to cart

Description

Description

This gene encodes the soluble mannose-binding lectin or mannose-binding protein found in serum. The protein encoded belongs to the collectin family and is an important element in the innate immune system. The protein recognizes mannose and N-acetylglucosamine on many microorganisms, and is capable of activating the classical complement pathway. Deficiencies of this gene have been associated with susceptibility to autoimmune and infectious diseases.
Specifications

Specifications

MBL2
Polyclonal
Unconjugated
MBL2
Liquid
MBL2
Synthetic peptide sequence (STSHLAVCEFPI) corresponding to the C-terminus amino acids of MBL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P11226
MBL2; man-binding lectin; MBL; MBP; MBP1; COLEC1; HSMBPC; mannose-binding lectin (protein C) 2, soluble (opsonic defect); man-binding protein; mannose binding protein; Mannose-binding lectin 2, soluble (opsonic defect); MGC116832; MGC116833; mannose-binding protein C; soluble mannose-binding lectin
Goat
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4153
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit