Learn More
anti-MCAM/CD146, APC, Clone: 128018, R&D Systems™
Manufacturer:  R&D Systems FAB932A025
 View more versions of this product
Catalog No. FAB932A025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MCAM/CD146
128018
Supplied in a saline solution containing BSA and Sodium Azide.
Purified
Mouse myeloma cell line NS0-derived recombinant human MCAM, Val24-Gly559
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
Detects human MCAM in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, rmOCAM, or rmMAdCAM-1.
Flow Cytometry
APC
AAA20824
Murine
IgG1
25 Tests
Primary
4162
Human