Learn More
anti-MCAM/CD146, PE, Clone: 128018, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB932P100
 View more versions of this product
Catalog No. FAB932P100
Add to cart
This item is not returnable. View return policy
Description
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
MCAM/CD146
128018
Flow Cytometry 10 uL/10^6 cells
AAA20824
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human MCAM in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm) OCAM, or rmMAdCAM-1.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
MCAM
Mouse myeloma cell line NS0-derived recombinant human MCAM Val24-Gly559 Accession # AAA20824
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4162
Human