Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM Mouse anti-Human, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611208

Catalog No. BDB611208


Add to cart

Description

Description

Tumor metastasis is determined by the metastatic capacity of subpopulations of tumor cells. The presence of cell adhesion molecules (CAMs) is a property of tumor cells that is essential for metastasis. MCAM (MUC18 or Mel-CAM) was identified on melanoma cells and is one of several CAMs that belong to the immunoglobulin superfamily. MCAM is an integral membrane glycoprotein that consists of five immunoglobulin-like domains, a transmembrane region, and a short cytoplasmic domain. The presence of several protein kinase recognition motifs in the cytoplasmic domain suggest a role for MCAM in signal transduction. MCAM is capable of both homotypic and heterotypic adhesion between cells, but its heterotypic ligand remains to be identified. It is sparsely expressed on benign melanocytic nevi and thin primary melanomas. However, MCAM expression increases with tumor thickness. It is expressed on 70-80% of advanced primary and metastatic tumors. Additionally, S-Endo-1 antigen (CD146), present on human endothelial cells, is identical to MCAM. Thus, MCAM may play an important role in the development of metastatic tumors and possibly represents a prime target for therapeutic treatment.

Host Species: Mouse
Clone: 1
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human MCAM aa. 104-305
Formula Weight [Chemical]: 113kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

MCAM
1
Unconjugated
Affinity Purified
Human MCAM aa. 104-305
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit