Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAR1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524283

Catalog No. PIPA524283


Add to cart

Specifications

Specifications

MCAR1
Polyclonal
PBS with 0.09% sodium azide
Q9H1U9, Q5HZI9
CG7943, mitochondrial carrier triple repeat protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230125, 92014
Western Blot
Unconjugated
SLC25A51
Liquid
SLC25A51
KLH conjugated synthetic peptide between 46-75 amino acids from the N-terminal region of human MCAR1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit