Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCC Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610740

Catalog No. BDB610740


Add to cart

Description

Description

The Mutated in Colorectal Cancer gene (MCC) was identified as a tumor suppressor that is deleted in sporadic and familial colorectal cancers. MCC is located at chromosome region 5q21. This region is often deleted in and associated with adenomas and carcinomas. Allele losses in the regions contiguous to MCC correlate with a high rate of colorectal tumors. MCC has been reported to be widely expressed and most abundant in epithelial cells and brain. It is a protein of 829 amino acids with a short sequence homologous to the m3 muscarinic acetylcholine receptor. Immunolocalization studies show MCC in the lateral plasma membrane of intestinal epithelial cells. In brain, MCC is abundant in the molecular layer of the cerebellum and is associated with various organelles, such as the plasma membrane. Although overexpression of wild type MCC protein blocks progression through the cell cycle, a mutated MCC, that contains a Gln instead of Arg-506, does not. Therefore, wild type MCC functions as a negative regulator of the cell cycle. Mutations or deletions in MCC might interfere with its physiological role and allow unrestricted cell proliferation (i.e. tumor formation).

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

MCC
1
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Mutated in Colorectal Cancer
Affinity Purified
Human MCC aa. 5-146
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit