Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCL1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121067

Catalog No. PIPA121067

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in programing of differentiation and concomitant maintenance of viability but not of proliferation. Isoform 1 inhibits apoptosis while isoform 2 promotes it.

PA1-21067 detects MCL1 in human samples.PA1-21067 has been successfully used in immunohistochemistry (paraffin) procedures.PA1-21067 immunogen is a synthetic peptide corresponding to residues S(121) P E E E L D G Y E P E P L G K R P A(139) of human MCL1.
Specifications

Specifications

MCL1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
Q8HYS5, Q7YRZ9, Q07820
BCL2L3, EAT, MCL1L, MCL1S, MGC104264, MGC1839, Mcl-1, TM
Rabbit
IgG
7mL
4° C, do not freeze
Primary
4170
Immunohistochemistry (Paraffin)
Unconjugated
MCL1
Purified
MCL1
Synthetic peptide corresponding to residues S(121) P E E E L D G Y E P E P L G K R P A(139) of human MCL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Feline, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit