Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MCM2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136085

Catalog No. NBP136085


Add to cart

Description

Description

MCM2 Polyclonal antibody specifically detects MCM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MCM2
Unconjugated
10mM PBS (pH 7.4) and 1.0% BSA with 0.1% Sodium Azide
BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin)
Rabbit
IgG
1 ml
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary
4171
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100
Affinity Purified
MCM2
Synthetic peptide corresponding to N-terminal of human MCM2 protein.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This product is specific for MCM2
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only