Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138099

Catalog No. 01-673-660


Add to cart

Description

Description

This antibody is predicted to react with bovine, canine, porcine and rabbit based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.

The mini-chromosome maintece (MCM) family of proteins, including MCM2, MCM3, MCM4(Cdc21), MCM5(Cdc46), MCM6(Mis5) and MCM7(Cdc47), are regulators of DNA replication that act to ensure replication occurs only once in the cell cycle. Expression of MCM proteins increases during cell growth, peaking at G1 to S phase. MCM2/MCM4 complexes function as substrates for Cdc2/cyclin B in vitro.
Specifications

Specifications

MCM2
Polyclonal
Unconjugated
MCM2
Liquid
MCM2
Synthetic peptide corresponding to N-terminus of human MCM2 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.75 mg/mL
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P97310, P49736
BM28, CCNL1, CDCL1, D3S3194, MITOTIN, cdc19, DNA replication licensing factor MCM2, MCM2 minichromosome maintece deficient 2, mitotin, cell devision cycle-like 1, cyclin-like 1, minichromosome maintece deficient 2 mitotin, minichromosome maintece protein 2 homolog, nuclear protein BM28
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4171, 17216
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit