Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7, Polyclonal, Invitrogen™

Catalog No. PIPA126585
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126585 50 μL
1 options

Catalog No. PIPA126585

Supplier: Thermo Scientific PA126585

Goat Polyclonal Antibody

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumorsuppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

PA1-26585 detects MCM7 from human samples. PA1-26585 is expected to cross react with mouse (78% conserved) due to sequence homology.PA1-26585 has been successfully used in Western blot procedures. Predicted molecular weight: 90 kDa. Positive control: HeLa cell lysate.The PA1-26585 immunogen is a synthetic peptide corresponding to a portion of human Minichromosome Maintenance Deficient 7 encoded in part by exons 8 and 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM7
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene MCM7
Gene Accession No. P33993, Q61881
Gene Alias CDC47, MCM2, P1.1-MCM3, P1CDC47, P85MCM, PNAS146
Gene Symbols MCM7
Host Species Goat
Immunogen Synthetic peptide corresponding to a portion of human Minichromosome Maintenance Deficient 7 encoded in part by exons 8 and 9.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4176
Target Species Human, Mouse
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.