Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM8, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131306

Catalog No. PIPA131306

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein contains the central domain that is conserved among the MCM proteins. This protein has been shown to co-immunoprecipitate with MCM4, 6 and 7, which suggests that it may interact with other MCM proteins and play a role in DNA replication. Alternatively spliced transcript variants encoding distinct isoforms have been described.

PA1-31306 detects MCM8 from human samples.PA1-31306 has been successfully used in immunoprecipitation, Western blot applications.The PA1-31306 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) representing a portion of MCM8 encoded within exon 2 (Human).
Specifications

Specifications

MCM8
Polyclonal
Unconjugated
MCM8
Purified
MCM8
Synthetic peptide corresponding to the representing a portion of MCM8 encoded within exon 2 (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunoprecipitation, Western Blot
1mg/mL
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8
Q9UJA3, Q9CWV1
C20orf154, MGC119522, MGC119523, MGC12866, MGC4816, REC, dJ967N21.5
Goat
IgG
100μL
Store at -20°C
Primary
84515
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit