Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14124

Catalog No. PA14124


Add to cart

Description

Description

PA1-4124 detects MCT-1 in mouse samples. PA1-4124 has been successfully used in Western blot procedure. By Western blot this antibody detects a ~20 kDa protein representing MCT-1 in transfected 3T3 cells. The PA1-4124 immunogen is a synthetic peptide corresponding to residues K(3) K F D E K E N V S N C(14) of the 20 kDa MCT-1.

SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Specifications

Specifications

MCT1
Polyclonal
Unconjugated
Slc16a1
Liquid
SLC16A1
Synthetic peptide corresponding to residues K(3) K F D E K E N V S N C(14) of the 20 kDa MCT-1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P53986
Multiple copies in a T-cell maligcies, Maligt T cell amplified sequence 1MCT1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
20501
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit