Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MD-1/LY86, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide corresponding to amino acids 112-125 of human MD-1.
Antigen MD-1/LY86
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056700T
View Documents
Novus Biologicals
NB10056700SS
0.025 mg
Each for $119.60
Add to cart
 
NB10056700
View Documents
Novus Biologicals
NB10056700
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

MD-1/LY86 Polyclonal antibody specifically detects MD-1/LY86 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
This antibody was developed against a KLH-conjugated synthetic peptide corresponding to amino acids 112-125 of human MD-1.
Polyclonal
Primary
Western Blot, Immunohistochemistry (Paraffin)
O95711
PBS with 0.05% Sodium Azide
IgG
MD-1/LY86
Purified
RUO
In Ramos, a 25 kDa band is observed.
9450
0.5 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit