Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520261
Description
A suggested positive control is Daudi cell lysate. PA5-20261 can be used with blocking peptide PEP-0376.
MD-1 Polyclonal antibody specifically detects MD-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MD-1 | |
Polyclonal | |
Unconjugated | |
LY86 | |
dJ80N2.1; Ly86; Ly-86; Lymphocyte antigen 86; MD1; MD-1; MD-1, RP105-associated; MMD-1; Protein MD-1; RP1-80N2.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17084, 291359, 9450 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O88188, O95711 | |
LY86 | |
A 15 amino acid peptide from near the center of human MD-1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction