Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MDC1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056657SS

 View more versions of this product

Catalog No. NB10056657T


Add to cart

Description

Description

MDC1 Polyclonal antibody specifically detects MDC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MDC1
0.5 mg/mL
Western Blot 1:200-1:1000
NP_055456
DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
Rabbit
IgG
0.05 ml
Autophagy, DNA Repair
Primary
9656
Human
Western Blot
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Purified
MDC1
This antibody was developed against a synthetic peptide corresponding to amino acids 871-888 (RQESDKNGESASPERDRE) of human NFBD1.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
In Jurkat, an approx. 220 kDa band is observed.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only