Learn More
mDIC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3840
Catalog No. PA3840
Add to cart
Description
Description
PA3-840 detects mitochondrial dicarboxylate carrier protein(mDIC) in human and mouse cells. PA3-840 has been successfully used in Western blotting and ICC/IF procedures. By Western blot it detects a 28 kDa protein representing mDIC. The PA3-840 immunogen is a synthetic peptide corresponding to the C-terminal residues L(276) R K H F G I K V P T T(287) of mDIC.
The mitochondrial dicarboxylate carrier protein (mDIC) belongs to superfamily of mitochondrial transporters. It has been demonstrated that the expression of mDIC leads to hyperpolarization of the mitochondria, as well as succinate uptake by the mitochondria. It is highly expressed in adipose tissue, where its expression is downregulated by insulin and upregulated by free fatty acids. mDIC is also thought to play a role in glyceroneogenesis.
Specifications
Specifications
mDIC
Polyclonal
whole serum with 0.05% sodium azide
Q9UBX3, Q9QZD8
Mitochondrial dicarboxylate carrier protein
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SLC25A10
Liquid
SLC25A10
Synthetic peptide corresponding to the C-terminal residues L(276) R K H F G I K V P T T(287) of mDIC.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1468, 27376