Learn More
MDH2 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP154649
Catalog No. NBP154649
Add to cart
Description
Description
MDH2 Polyclonal antibody specifically detects MDH2 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
MDH2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
MDH2
Synthetic peptides corresponding to MDH2(malate dehydrogenase 2, NAD (mitochondrial)) The peptide sequence was selected from the C terminal of MDH2 (NP_005909). Peptide sequence TYFSTPLLLGKKGIEKNLGIGKVSSFEEKMISDAIPELKASIKKGEDFVK.
33 kDa
100ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by.
Immunohistochemistry, Western Blot
Western Blot 2.5 ug/ml, Immunohistochemistry
P40926
EC 1.1.1, EC 1.1.1.37, malate dehydrogenase 2, NAD (mitochondrial), MDH, MGC:3559, mitochondrial, MOR1
Rabbit
IgG
Protein A purified
RUO
Primary
4191
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish