Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2 Mouse anti-Human, Mouse, Clone: 3G9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041530
Description
Anti-MDM2 Antibody, clone 3G9 is a Mouse Monoclonal Antibody for detection of MDM2 also known as Double minute 2 protein or Mdm2 p53 binding protein homolog and has been validated in western blot, immunoprecipitation, immunocytochemistry and immunohistochemistry.
Specifications
MDM2 | |
Monoclonal | |
Unconjugated | |
NP_002383.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
3G9 | |
Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
MDM2, hdm2, HDMX, Hdm2, HDM2 | |
His-tagged recombinant protein corresponding to human MDM2. | |
100 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction