Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDMX, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528212

Catalog No. PIPA528212

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The human MDM4 gene, which plays a role in apoptosis, encodes a 490-amino acid protein containing a RING finger domain and a putative nuclear localization signal. The MDM4 putative nuclear localization signal, which all Mdm proteins contain, is located in the C-terminal region of the protein. The mRNA is expressed at a high level in thymus and at lower levels in all other tissues tested. MDM4 protein produced by in vitro translation interacts with p53 via a binding domain located in the N-terminal region of the MDM4 protein. MDM4 shows significant structural similarity to p53-binding protein MDM2. Two transcript variants, one protein-coding and the other likely not to be protein-coding, have been found for this gene.

PA5-28212 targets MDMX in IF, IHC (P), and WB applications and shows reactivity with Human samples.The PA5-28212 immunogen is recombinant protein fragment corresponding to a region within amino acids 62 and 490 of Human MDMX.
Specifications

Specifications

MDMX
Polyclonal
Unconjugated
MDM4
Purified
MDM4
Recombinant protein fragment corresponding to a region within amino acids 62 and 490 of Human MDMX
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
4194
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O15151
HDMX, MDMX, MRP1, MDM4-related protein 1, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, double minute 4, human homolog of, p53-binding protein, mdm2-like p53-binding protein, protein Mdm4, protein Mdmx
Rabbit
IgG
100μL
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit