anti-MDR1, Conformational Extracellular Epitope, Clone: UIC2, Millipore™ (Chemicon™)

Manufacturer: EMD Millipore MAB4334Z/DEL

 View more versions of this product

Catalog No. 50-175-512

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Antigen MDR1, Conformational Extracellular Epitope
    Format Semi-purified
    Host Species Mouse
    Isotype IgG2a κ
    Regulatory Status RUO
    Cross Reactivity Human
    Immunogen Mouse BALB/c 3T3 fibroblasts transfected with human MDR1 cDNA, followed by a multistep selection of tranfectants for resistance to vinblastine.
    Quantity 100μg
    Research Discipline Endocrine/Metabolism
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Conjugate Unlabeled
    Chains or Fragments P-glycoprotein encoded by the MDR1 gene (ABCB1; LocusLink ID: 5243)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit