Learn More
Please call 1-800-766-7000 to place your order or try our site again later.
MilliporeSigma anti-Measles Blend, hemagglutinin, Clone: CV1, CV4, (Chemicon)
Manufacturer: MilliporeSigma MAB8905/DEL
Description
Immunofluorescence, Immunohistochemistry
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Measles Blend, hemagglutinin | |
Ascites | |
Hemagglutinin protein obtained from Chicago wild type strain of measles | |
100μL | |
Primary | |
Human |
Unconjugated | |
Murine | |
IgG1 | |
A blend of two monoclonal antibodies that recognize the measles hemagglutinin protein by indirect immunofluorescence. | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.