Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MECP2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127874

Catalog No. PIPA127874

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Chromosomal protein that binds to methylated DNA. It can bind specifically to a single methyl-CpG pair. It is not influenced by sequences flanking the methyl-CpGs. Mediates transcriptional repression through interaction with histone deacetylase and the co

PA1-27874 detects MECP2 from human, mouse, and rat samples.PA1-27874 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Methyl-CpG-binding protein 2.
Specifications

Specifications

MECP2
Polyclonal
PBS with no preservative
P51608, Q9Z2D6, Q00566
AUTSX3, DKFZp686A24160, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
Chicken
IgY
50μg
Store at -20°C
Primary
4204
Western Blot
Unconjugated
MECP2
Purified
MECP2
Synthetic peptide corresponding to human Methyl-CpG-binding protein 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit