Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED25 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523711

Catalog No. PIPA523711

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

This gene encodes a component of the transcriptional coactivator complex termed the Mediator complex. This complex is required for transcription of most RNA polymerase II-dependent genes. The encoded protein plays a role in chromatin modification and in preinitiation complex assembly. Mutations in this gene are associated with Charcot-Marie-Tooth disease type 2B2.
Specifications

Specifications

MED25
Polyclonal
PBS with 0.09% sodium azide
Q71SY5, Q8VCB2
ACID1, ARC92, CMT2B2, P78, PTOV2, ARC/mediator transcriptional coactivator subunit, activator interaction domain-containing protein 1, activator-recruited cofactor 92 kDa component, mediator of RNA polymerase II transcription subunit 25, mediator of RNA polymerase II transcription, subunit 25
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
75613, 81857
Immunohistochemistry, Western Blot
Unconjugated
MED25
Liquid
MED25
KLH conjugated synthetic peptide between 112-140 amino acids from the N-terminal region of human MED25
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit