Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138111

Catalog No. 01-673-666

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The myocyte enhancer factors-2 (MEF-2) family of transcription factors associate with a variety of co-repressors or co-activators to regulate development and function of T cells, neuronal cells, and muscle cells. Four distinct family members arise from alternatively splices transcripts, termed MEF-2A, MEF-2B, MEF-2C, and MEF-2D. These members bind as homo- and heterodimers to the MEF2 site in the promoter region of affected genes. Differential regulation in the expression of the 4 transcripts implies functional distinction for each during embryogenesis and development.

This antibody is predicted to react with bovine, chicken, canine, amphibian, mouse, porcine, rabbit and rat based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is brain tissue.
Specifications

Specifications

MEF2A
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
Q02078
ADCAD1, RSRFC4, RSRFC9, mef2, MADS box transcription enhancer factor 2, polypeptide A myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, serum response factor-like protein 1
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4205
Immunohistochemistry (Paraffin)
Unconjugated
MEF2A
Liquid
MEF2A
Synthetic peptide corresponding to aa 487-507 of human MEF-2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit