Learn More
MEF2A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526266
Catalog No. PIPA526266
Add to cart
Description
Description
This antibody is predicted to react with bovine, porcine and rat based on sequence homology.
MEF2A is a DNA-binding transcription factor that activates many muscle-specific, growth factor-induced, and stress-induced genes. The encoded protein can act as a homodimer or as a heterodimer and is involved in several cellular processes, including muscle development, neuronal differentiation, cell growth control, and apoptosis. Defects in this protein could be a cause of autosomal domit coronary artery disease 1 with myocardial infarction (ADCAD1).
Specifications
Specifications
MEF2A
Polyclonal
Unconjugated
MEF2A
Liquid
MEF2A
KLH conjugated synthetic peptide between 453-479 amino acids from the central region of human MEF2A
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide
Q02078, Q60929
ADCAD1, RSRFC4, RSRFC9, mef2, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A), myocyte-specific enhancer factor 2A, serum response factor-like protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17258, 4205