Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513289

Catalog No. PIPA513289

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, mouse, porcine, rat and Xenopus based on sequence homology.

MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. This protein controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. It may also be involved in neurogenesis and in the development of cortical architecture.
Specifications

Specifications

MEF2C
Polyclonal
PBS with 0.09% sodium azide
Q06413
Myocyte-specific enhancer factor 2C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4208
Western Blot
Unconjugated
MEF2C
Liquid
MEF2C
KLH conjugated synthetic peptide between 37-66 amino acids from human MEF2C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit