Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2D Mouse, Unlabeled, Clone: 9, BD

Mouse Monoclonal Antibody

$225.00 - $445.00

Specifications

Antigen MEF2D
Isotype IgG1
Immunogen Mouse MEF2D aa. 346-511
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610775
View Documents
BD Biosciences
610775
150μg
Each for $445.00
Add to cart
 
BDB610774
View Documents
BD Biosciences
610774
50μg
Each for $225.00
Add to cart
 
Description

Description

MEF2D is a member of the MEF2 (Myocyte-specific Enhancer-binding Factor 2) transcription factor family. Members of this family (MEF2A, B, C, & D) contain several characteristic motifs that include a MEF2 domain and a MADS box. MEF2 proteins bind the DNA sequence, 5 ft.- (C/T)T(A/T)(A/T)AAATA(A/G)-3 ft., and binding occurs primarily, although not exclusively, in muscle cells. MEF2D gene expression is ubiquitous and the mRNA is alternatively spliced. This results in multiple protein isoforms that heterodimerize with other family members. In addition to the MEF2 and MADS domains, MEF2D has a glutamine-/proline-rich region, amino acids 365-404, and a basic region, in the MADS domain near the N-terminus. The glutamine-/proline-rich region is involved in transactivation, while the basic region mediates sequence-specific DNA binding. MEF2D displays 75% similarity to hMEF2A and hMEF2C. Myogenin gene transcription requires MEF2D, indicating that MEF2D is involved in myogenesis and muscle cell differentiation. MEF2D has been reported to have a calculated molecular weight of 56kDa, but can be observed migrating in a range of 56-70kDa.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

MEF2D
Mouse MEF2D aa. 346-511
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Myocyte-specific Enhancer-binding Factor 2
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
9
250μg/mL
Store undiluted at -20°C.
SDS

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit