Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEFV Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528397

Catalog No. PIPA528397


Add to cart

Description

Description

PA5-28397 targets MEFV in IF and WB applications and shows reactivity with Human samples. The PA5-28397 immunogen is synthetic peptide corresponding to a region within amino acids 378 and 470 of Human MEFV. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein, also known as pyrin or marenostrin, that is an important modulator of innate immunity. Mutations in this gene are associated with Mediterranean fever, a hereditary periodic fever syndrome.
Specifications

Specifications

MEFV
Polyclonal
Unconjugated
MEFV
Liquid
MEFV
Synthetic peptide corresponding to a region within amino acids 378 and 470 of Human MEFV
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.67 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
O15553
FMF, MEF, TRIM20, marenostrin, pyrin
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4210
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit