Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEIG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521156
Description
A suggested positive control is K562 cell lysate. PA5-21156 can be used with blocking peptide PEP-1270.
MEIG1 Polyclonal antibody specifically detects MEIG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MEIG1 | |
Polyclonal | |
Unconjugated | |
MEIG1 | |
bA2K17.3; Meg1; Meig1; meiosis expressed gene 1; meiosis expressed gene 1 homolog; meiosis expressed gene 1 protein homolog; meiosis/spermiogenesis associated 1; Meiosis-expressed gene 1 protein; MLZ-278; Protein expressed in male leptotene and zygotene spermatocytes 278; SPATA39; spermatogenesis associated 39 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
104362, 644890, 679834 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5JSS6, Q61845 | |
MEIG1 | |
A 20 amino acid peptide near the carboxy terminal of human MEIG1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction