Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MEIS1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224522SS

 View more versions of this product

Catalog No. NBP224522SS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MEIS1 Polyclonal antibody specifically detects MEIS1 in Human, Mouse, Rat, Canine, Chicken, Equine, Xenopus samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

MEIS1
0.5 mg/mL
Immunohistochemistry-Paraffin 5 ug/ml
O00470
homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse)
Rabbit
IgG
0.025 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, rat, mouse, monkey, cow, and dog, and 93% homologous in horse.
Immunohistochemistry (Paraffin)
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Affinity Purified
MEIS1
A portion of amino acids 280-330 of MEIS1 was used as the immunogen.
Immunogen affinity purified
RUO
Primary
4211
Human, Mouse, Rat, Canine, Chicken, Equine, Xenopus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only