Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIS1 Rabbit anti-Canine, Chicken, Equine, Human, Mouse, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523377

Catalog No. PIPA523377


Add to cart

Description

Description

MEIS1, also known as Homeobox protein Meis1, is a nuclear protein belonging to the TALE/MEIS homeobox family with a homeobox DNA-binding domain. It acts as a transcriptional regulator of PAX6 (Paired box protein Pax-6). It is a protein required for hematopoiesis, megakaryocyte lineage development and vascular patterning. Expression is seen at low levels in normal immunohepatopoietic tissues, and in a subset of myeloid leukemia cell lines, with the highest expression seen in those with a megakaryocytic-erythroid phenotype. It is also expressed at high levels in the cerebellum.
Specifications

Specifications

MEIS1
Polyclonal
Unconjugated
MEIS1
Liquid
MEIS1, meis1.S
Synthetic peptide (296 THPYPSEEQKKQLAQD 311) of MEIS1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Chicken, Equine, Human, Mouse, Rat, Xenopus laevis
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O00470, Q60954, P79937
Meis1, myeloid ecotropic viral integration site 1 homolog, WUGSC:H_NH0444B04.1, homeobox protein Meis1, leukemogenic homolog protein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100051257, 17268, 395589, 399305, 4211, 474619, 686117
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit